accessibility menu, dialog, popup

UserName

CAS RN 786616-54-4

NH2FOOH

CAS RN 786616-54-4

IUPAC Name: 3-amino-5-fluorobenzoic acid
Synonyms: 3-amino-5-fluorobenzoic acid
Molecular Weight (g/mol): 155.13
Molecular Formula: C7H6FNO2
InChi Key: RYLBYCHERDTVAY-UHFFFAOYSA-N
SMILES: NC1=CC(F)=CC(=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results