CAS RN 78821-43-9
CAS RN 78821-43-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Epibrassinolide, MedChemExpress
MedChemExpress Epibrassinolide (24-Epibrassinolide) is a ubiquitously occurring plant growth hormone which shows great potential to alleviate heavy metals and pesticide stress in plants. Epibrassinolide is a potential apoptotic inducer in various cancer cells without affecting the non-tumor cell growth.