accessibility menu, dialog, popup

UserName

CH3H2NOH

CAS RN 83647-42-1

IUPAC Name: (3-amino-2-methylphenyl)methanol
Synonyms: (3-amino-2-methylphenyl)methanol
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: UVYZMJMDIMWDNJ-UHFFFAOYSA-N
SMILES: CC1=C(CO)C=CC=C1N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results