CAS RN 844499-71-4
CAS RN 844499-71-4
IUPAC Name:
4-hydroxy-3-{2'-hydroxy-[1,1'-biphenyl]-4-yl}-6-oxo-6H,7H-thieno[2,3-b]pyridine-5-carbonitrile
Synonyms:
4-hydroxy-3-{2'-hydroxy-[1,1'-biphenyl]-4-yl}-6-oxo-7H-thieno[2,3-b]pyridine-5-carbonitrile
Molecular Weight (g/mol):
360.39
Molecular Formula:
C20H12N2O3S
InChi Key:
CTESJDQKVOEUOY-UHFFFAOYSA-N
SMILES:
OC1=CC=CC=C1C1=CC=C(C=C1)C1=CSC2=C1C(O)=C(C#N)C(=O)N2
1
–
3
of
3
results
1
Tocris Bioscience™ A 769662
CAS: 844499-71-4 Molecular Formula: C20H12N2O3S
2
A-769662, MedChemExpress
MedChemExpress A-769662 is a potent, reversible AMPK activator with EC50 of 0.8 μM.
3