CAS RN 84803-56-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-(2-Methoxyethyl)aniline, TRC
CAS: 84803-56-5 Molecular Formula: C9H13NO Molecular Weight (g/mol): 151.21 Synonym: 4-(2-Methoxyethyl)benzenamine,p-2-Methoxyethyl-Aniline,1-Methoxy-2-(4-aminophenyl)ethane,4-(2-Methoxyethyl)phenylamine,p-(2-Methoxyethyl)aniline IUPAC Name: 4-(2-methoxyethyl)aniline SMILES: COCCc1ccc(N)cc1