CAS RN 850375-12-1
IUPAC Name:
2-bromo-1-(5-bromo-1-benzothiophen-3-yl)ethan-1-one
Synonyms:
2-bromo-1-(5-bromo-1-benzothiophen-3-yl)ethanone
Molecular Weight (g/mol):
334.03
Molecular Formula:
C10H6Br2OS
InChi Key:
QFZPWQQHQRYUKB-UHFFFAOYSA-N
SMILES:
BrCC(=O)C1=CSC2=CC=C(Br)C=C12