accessibility menu, dialog, popup

UserName

CH3CH3CMgBr(E/Z)

CAS RN 85676-85-3

IUPAC Name: magnesium(2+) but-2-en-2-ide bromide
Synonyms: magnesium(2+) but-2-en-2-ide bromide
Molecular Weight (g/mol): 159.31
Molecular Formula: C4H7BrMg
InChi Key: FGYXQBPXHQHYNU-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].CC=[C-]C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results