accessibility menu, dialog, popup

UserName

CAS RN 86225-73-2

IUPAC Name: 3-chloro-2,6-difluorobenzonitrile
Synonyms: 3-chloro-2,6-difluorobenzonitrile
Molecular Weight (g/mol): 173.55
Molecular Formula: C7H2ClF2N
InChi Key: QYALCKFGISNFMJ-UHFFFAOYSA-N
SMILES: FC1=CC=C(Cl)C(F)=C1C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results