missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2,6-difluorobenzonitrile, 97%, Thermo Scientific™

Product Code. 11491220
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11491220 5 g 5g
11481220 1 g 1g
11401230 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491220 Supplier Thermo Scientific Alfa Aesar Supplier No. H26335.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Chloro-2,6-difluorobenzonitrile is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86225-73-2
Molecular Formula C7H2ClF2N
Molecular Weight (g/mol) 173.55
MDL Number MFCD06660172
InChI Key QYALCKFGISNFMJ-UHFFFAOYSA-N
Synonym benzonitrile, 3-chloro-2,6-difluoro, ksc495c2b, 3-chloro-2,6-difluoroben, 3-chloro-2,6-difluorobenzenecarbonitrile
PubChem CID 13175231
IUPAC Name 3-chloro-2,6-difluorobenzonitrile
SMILES FC1=CC=C(Cl)C(F)=C1C#N

Specifications

Quantity 5 g
Refractive Index 1.5125
UN Number UN3276
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 173.55
Percent Purity 97%
Chemical Name or Material 3-Chloro-2,6-difluorobenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.