accessibility menu, dialog, popup

UserName

KCH3O

CAS RN 865-33-8

IUPAC Name: potassium methanolate
Synonyms: potassium methoxide
Molecular Weight (g/mol): 70.13
Molecular Formula: CH3KO
InChi Key: BDAWXSQJJCIFIK-UHFFFAOYSA-N
SMILES: [K+].C[O-]
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results