CAS RN 866206-54-4
CAS RN 866206-54-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
PRX-08066, MedChemExpress
MedChemExpress PRX-08066 is a selective 5-hydroxytryptamine receptor 2B (5-HT2BR, IC50= 3.4 nM) antagonist that causes selective vasodilation of pulmonary arteries.