accessibility menu, dialog, popup

UserName

CH3H3CONBr

CAS RN 870521-31-6

IUPAC Name: 5-bromo-2-(propan-2-yloxy)pyridine
Synonyms: 5-bromo-2-isopropoxypyridine
Molecular Weight (g/mol): 216.08
Molecular Formula: C8H10BrNO
InChi Key: FSZRSTNRZUIEMA-UHFFFAOYSA-N
SMILES: CC(C)OC1=NC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results