accessibility menu, dialog, popup

UserName

CAS RN 870563-60-3

H3COFNH2

CAS RN 870563-60-3

IUPAC Name: 1-(4-fluoro-2-methoxyphenyl)methanamine
Synonyms: 1-(4-fluoro-2-methoxyphenyl)methanamine
Molecular Weight (g/mol): 155.17
Molecular Formula: C8H10FNO
InChi Key: OUYQGZMPYJPPER-UHFFFAOYSA-N
SMILES: COC1=C(CN)C=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results