CAS RN 871361-88-5
IUPAC Name:
2,6-bis[(pyridin-4-yl)methylidene]cyclohexan-1-one
Synonyms:
2,6-bis(pyridin-4-ylmethylidene)cyclohexan-1-one
Molecular Weight (g/mol):
276.34
Molecular Formula:
C18H16N2O
InChi Key:
CYVVJSKZRBZHAV-UHFFFAOYSA-N
SMILES:
O=C1C(CCCC1=CC1=CC=NC=C1)=CC1=CC=NC=C1
1
–
1
of
1
results
1
SC66, MedChemExpress
MedChemExpress SC66 is an Akt inhibitor, reduces cell viability in a dose- and time-dependent manner, inhibits colony formation and induces apoptosis in hepatocellular carcinoma (HCC) cells.