accessibility menu, dialog, popup

UserName

CAS RN 87151-66-4

HO

CAS RN 87151-66-4

IUPAC Name: 3-(cyclohexa-1,4-dien-1-yl)propan-1-ol
Synonyms: 3-(cyclohexa-1,4-dien-1-yl)propan-1-ol
Molecular Weight (g/mol): 138.21
Molecular Formula: C9H14O
InChi Key: DJJNAQALDCPMOU-UHFFFAOYSA-N
SMILES: OCCCC1=CCC=CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results