accessibility menu, dialog, popup

UserName

CH3CH3H3CSiCH3H3COHO

CAS RN 87184-99-4

IUPAC Name: 4-[(tert-butyldimethylsilyl)oxy]butan-1-ol
Synonyms: 4-[(tert-butyldimethylsilyl)oxy]butan-1-ol
Molecular Weight (g/mol): 204.39
Molecular Formula: C10H24O2Si
InChi Key: IJEMXJANZPVITP-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](C)(C)OCCCCO
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)