CAS RN 87269-87-2
CAS RN 87269-87-2
IUPAC Name:
benzyl octahydrocyclopenta[b]pyrrole-2-carboxylate hydrochloride
Synonyms:
benzyl octahydrocyclopenta[b]pyrrole-2-carboxylate hydrochloride
Molecular Weight (g/mol):
281.78
Molecular Formula:
C15H20ClNO2
InChi Key:
HLXCXOQXUDRJLF-UHFFFAOYNA-N
SMILES:
Cl.O=C(OCC1=CC=CC=C1)C1CC2CCCC2N1
1
–
4
of
4
results
1
(2S,3aS,6aS)-Octahydrocyclopenta[b]pyrrole-2-carboxylic Acid Phenylmethyl Ester Hydrochloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
(S,S,S)-2-Azabicyclo[3.3.0]octane-3-carboxylic Acid, Benzyl Ester, Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(S,S,S)-2-Azabicyclo[3.3.0]octane-3-carboxylic Acid, Benzyl Ester, Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
(S,S,S)-2-Azabicyclo[3.3.0]octane-3-carboxylic Acid, Benzyl Ester, Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.