CAS RN 87453-76-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
3-Dimethylaminopropyl-2-benzylaminobenzoate, TRC
CAS: 87453-76-7 Molecular Formula: C19 H24 N2 O2 Molecular Weight (g/mol): 312.41 Synonym: 2-[(Phenylmethyl)amino]benzoic acid 3-(Dimethylamino)propyl ester,Benzoic acid, 2-[(phenylmethyl)amino]-, 3-(dimethylamino)propyl ester,3-(Dimethylamino)propyl 2-[(phenylmethyl)amino]benzoate,3-Dimethylaminopropyl-2-benzylaminobenzoate IUPAC Name: 3-(dimethylamino)propyl 2-(benzylamino)benzoate SMILES: CN(C)CCCOC(=O)c1ccccc1NCc2ccccc2
2
3-(Dimethylamino)propyl-2-(benzylamino)benzoate, Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.