Learn More
3-Dimethylaminopropyl-2-benzylaminobenzoate, TRC
Specifications
Specifications
| Quantity | 1 g |
| InChI Formula | InChI=1S/C19H24N2O2/c1-21(2)13-8-14-23-19(22)17-11-6-7-12-18(17)20-15-16-9-4-3-5-10-16/h3-7,9-12,20H,8,13-15H2,1-2H3 |
| Formula Weight | 312.1838 |
| Percent Purity | >95% |
| Chemical Name or Material | 3-Dimethylaminopropyl-2-benzylaminobenzoate |
| Grade | HPLC |