CAS RN 87460-09-1
CAS RN 87460-09-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
Benzyl Hydroxy(4-phenylbutyl)phosphinoacetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Benzyl Hydroxy(4-phenylbutyl)phosphinoacetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
[2-(Benzyloxy)-2-oxoethyl](4-phenylbutyl)phosphinic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4
Benzyl Hydroxy(4-phenylbutyl)phosphinoacetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.