accessibility menu, dialog, popup

UserName

H3COOHH3C

CAS RN 875-59-2

IUPAC Name: 1-(4-hydroxy-2-methylphenyl)ethan-1-one
Synonyms: 3-methyl-4-acetylphenol
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: IAMNVCJECQWBLZ-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(O)C=C1C
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results