accessibility menu, dialog, popup

UserName

CAS RN 875258-85-8

H3CNOFONNH3CCH3HCl

CAS RN 875258-85-8

IUPAC Name: 6-(4-fluorophenoxy)-2-methyl-3-{[1-(propan-2-yl)piperidin-3-yl]methyl}-3,4-dihydroquinazolin-4-one hydrochloride
Synonyms: 6-(4-fluorophenoxy)-3-[(1-isopropylpiperidin-3-yl)methyl]-2-methylquinazolin-4-one hydrochloride
Molecular Weight (g/mol): 445.96
Molecular Formula: C24H29ClFN3O2
InChi Key: QQGRMONBDBMLSP-UHFFFAOYNA-N
SMILES: Cl.CC(C)N1CCCC(CN2C(C)=NC3=CC=C(OC4=CC=C(F)C=C4)C=C3C2=O)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results