CAS RN 87579-78-0
CAS RN 87579-78-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
(2S,5R)-3,3-dimethyl-7-oxo-4-oxide-4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid Diphenylmethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(2S,5R)-3,3-dimethyl-7-oxo-4-oxide-4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid Diphenylmethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(2S,5R)-3,3-dimethyl-7-oxo-4-oxide-4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid Diphenylmethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.