CAS RN 87625-92-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Methyl 2-Methyl-6-(3-nitrophenyl)-4-oxo-2-cyclohexene-1-carboxylate + ~8% Ethyl 3-Methyl-3'-nitro-5-oxo-1,2,5,6-tetrahydro-[1,1'-biphenyl]-2-carboxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Methyl 2-Methyl-6-(3-nitrophenyl)-4-oxo-2-cyclohexene-1-carboxylate + ~8% Ethyl 3-Methyl-3'-nitro-5-oxo-1,2,5,6-tetrahydro-[1,1'-biphenyl]-2-carboxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Methyl 2-Methyl-6-(3-nitrophenyl)-4-oxo-2-cyclohexene-1-carboxylate, TRC
CAS: 87625-92-1 Molecular Formula: C15H15NO5 . C 16H17NO5 Molecular Weight (g/mol): 592.59 Synonym: Methyl 2-Methyl-6-(3-nitrophenyl)-4-oxo-2-cyclohexene-1-carboxylic Acid Ester IUPAC Name: methyl 2-methyl-6-(3-nitrophenyl)-4-oxo-cyclohex-2-ene-1-carboxylate SMILES: O=C1CC(C2=CC([N+]([O-])=O)=CC=C2)C(C(OC)=O)C(C)=C1.O=C3CC(C4=CC([N+]([O-])=O)=CC=C4)C(C(OCC)=O)C(C)=C3