missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Methyl-6-(3-nitrophenyl)-4-oxo-2-cyclohexene-1-carboxylate, TRC

Product Code. 30134656
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134656 250 mg 250mg
30004518 25 mg 25mg
30009953 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134656 Supplier Toronto Research Chemicals Supplier No. TRCM338310250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87625-92-1
Molecular Formula C15H15NO5 . C 16H17NO5
Molecular Weight (g/mol) 592.59
Synonym Methyl 2-Methyl-6-(3-nitrophenyl)-4-oxo-2-cyclohexene-1-carboxylic Acid Ester
IUPAC Name methyl 2-methyl-6-(3-nitrophenyl)-4-oxo-cyclohex-2-ene-1-carboxylate
SMILES O=C1CC(C2=CC([N+]([O-])=O)=CC=C2)C(C(OC)=O)C(C)=C1.O=C3CC(C4=CC([N+]([O-])=O)=CC=C4)C(C(OCC)=O)C(C)=C3

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C15H15NO5/c1-9-6-12(17)8-13(14(9)15(18)21-2)10-4-3-5-11(7-10)16(19)20/h3-7,13-14H,8H2,1-2H3
Formula Weight 592.21
Percent Purity >95%
Chemical Name or Material Methyl 2-Methyl-6-(3-nitrophenyl)-4-oxo-2-cyclohexene-1-carboxylate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.