CAS RN 876728-43-7
IUPAC Name:
(9H-fluoren-9-yl)methyl 4-(but-3-enoyl)piperazine-1-carboxylate
Synonyms:
9H-fluoren-9-ylmethyl 4-(but-3-enoyl)piperazine-1-carboxylate
Molecular Weight (g/mol):
376.46
Molecular Formula:
C23H24N2O3
InChi Key:
WRSRYPVHNSMIQR-UHFFFAOYSA-N
SMILES:
C=CCC(=O)N1CCN(CC1)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12