CAS RN 87687-02-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Resorufin Benzyl Ether, TRC
CAS: 87687-02-3 Molecular Formula: C19H13NO3 Molecular Weight (g/mol): 303.31 Synonym: 7-(Phenylmethoxy)-3H-phenoxazin-3-one,7-Benzyloxyphenoxazone,7-Benzyloxyresorufin IUPAC Name: 7-(benzyloxy)-3H-phenoxazin-3-one SMILES: O=C1C=CC2=NC3=C(OC2=C1)C=C(OCC4=CC=CC=C4)C=C3