Learn More
Resorufin Benzyl Ether, TRC
Product Code. 30428741
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30428741 5 mg 5mg
30416213 10 mg 10mg
30415591 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30428741 Supplier Toronto Research Chemicals Supplier No. TRCR1465185MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 87687-02-3
Molecular Formula C19H13NO3
Molecular Weight (g/mol) 303.31
Synonym 7-(Phenylmethoxy)-3H-phenoxazin-3-one, 7-Benzyloxyphenoxazone, 7-Benzyloxyresorufin
IUPAC Name 7-(benzyloxy)-3H-phenoxazin-3-one
SMILES O=C1C=CC2=NC3=C(OC2=C1)C=C(OCC4=CC=CC=C4)C=C3
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C19H13NO3/c21-14-6-8-16-18(10-14)23-19-11-15(7-9-17(19)20-16)22-12-13-4-2-1-3-5-13/h1-11H,12H2
Formula Weight 303.09
Chemical Name or Material Resorufin Benzyl Ether