CAS RN 877-11-2
IUPAC Name:
1,2,3,4,5-pentachloro-6-methylbenzene
Synonyms:
pentachlorotoluene
Molecular Weight (g/mol):
264.35
Molecular Formula:
C7H3Cl5
InChi Key:
AVSIMRGRHWKCAY-UHFFFAOYSA-N
SMILES:
CC1=C(Cl)C(Cl)=C(Cl)C(Cl)=C1Cl
1
–
2
of
2
results
1
2,3,4,5,6-Pentachlorotoluene, TRC
CAS: 877-11-2 Molecular Formula: C7 H3 Cl5 Molecular Weight (g/mol): 264.36 Synonym: Benzene, pentachloromethyl- (9CI),Toluene, 2,3,4,5,6-pentachloro- (6CI,7CI,8CI),1,2,3,4,5-Pentachloro-6-methylbenzene,2,3,4,5,6-Pentachlorotoluene,Methylpentachlorobenzene,NSC 16046 IUPAC Name: 1,2,3,4,5-pentachloro-6-methylbenzene SMILES: Cc1c(Cl)c(Cl)c(Cl)c(Cl)c1Cl