missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentachlorotoluene, TRC

Product Code. 17994455
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17994455 50 mg 50mg
17980445 100 mg 100mg
17960905 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17994455 Supplier Toronto Research Chemicals Supplier No. TRCP87711050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 877-11-2
Molecular Formula C7 H3 Cl5
Molecular Weight (g/mol) 264.36
Synonym Benzene, pentachloromethyl- (9CI), Toluene, 2,3,4,5,6-pentachloro- (6CI,7CI,8CI), 1,2,3,4,5-Pentachloro-6-methylbenzene, 2,3,4,5,6-Pentachlorotoluene, Methylpentachlorobenzene, NSC 16046
IUPAC Name 1,2,3,4,5-pentachloro-6-methylbenzene
SMILES Cc1c(Cl)c(Cl)c(Cl)c(Cl)c1Cl

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C7H3Cl5/c1-2-3(8)5(10)7(12)6(11)4(2)9/h1H3
Formula Weight 261.8677
Percent Purity >95%
Chemical Name or Material 2,3,4,5,6-Pentachlorotoluene
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.