CAS RN 87802-54-8
CAS RN 87802-54-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Pyridinamine-d4, TRC
CAS: 87802-54-8 Molecular Formula: C5H2D4N2 Molecular Weight (g/mol): 98.14 Synonym: 2-Aminopyridine-d4,1,2-Dihydro-2-iminopyridine-d4,2-Aminopyridine-d4,2-Pyridinylamine-d4,2-Pyridylamine-d4,NSC 431-d4,o-Aminopyridine-d4,α-Aminopyridine-d4,α-Pyridinamine-d4,α-Pyridylamine-d4 SMILES: [2H]C1=C([2H])C([2H])=C([2H])C(N)=N1