Learn More
2-Pyridinamine-d4, TRC
Product Code. 30424739
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424739 25 mg -
30410808 50 mg 50mg
30422573 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30424739 Supplier Toronto Research Chemicals Supplier No. TRCP99178725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 87802-54-8
Molecular Formula C5H2D4N2
Molecular Weight (g/mol) 98.14
Synonym 2-Aminopyridine-d4, 1,2-Dihydro-2-iminopyridine-d4, 2-Aminopyridine-d4, 2-Pyridinylamine-d4, 2-Pyridylamine-d4, NSC 431-d4, o-Aminopyridine-d4, α-Aminopyridine-d4, α-Pyridinamine-d4, α-Pyridylamine-d4
SMILES [2H]C1=C([2H])C([2H])=C([2H])C(N)=N1
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C5H6N2/c6-5-3-1-2-4-7-5/h1-4H,(H2,6,7)/i1D,2D,3D,4D
Formula Weight 98.0782
Chemical Name or Material 2-Pyridinamine-d4