accessibility menu, dialog, popup

UserName

H3CCH3OOOOCH3CH3

CAS RN 87826-26-4

IUPAC Name: 1,2-bis(8-methylnonyl) cyclohex-4-ene-1,2-dicarboxylate
Synonyms: 1,2-bis(8-methylnonyl) cyclohex-4-ene-1,2-dicarboxylate
Molecular Weight (g/mol): 450.7
Molecular Formula: C28H50O4
InChi Key: XRFYGUOAWVKOCQ-UHFFFAOYNA-N
SMILES: CC(C)CCCCCCCOC(=O)C1CC=CCC1C(=O)OCCCCCCCC(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results