accessibility menu, dialog, popup

UserName

CAS RN 878572-08-8

H3CFOCH3OH

CAS RN 878572-08-8

IUPAC Name: 1-(5-fluoro-2-methoxyphenyl)ethan-1-ol
Synonyms: 1-(5-fluoro-2-methoxyphenyl)ethanol
Molecular Weight (g/mol): 170.18
Molecular Formula: C9H11FO2
InChi Key: VLBOHPCLXJIASU-UHFFFAOYNA-N
SMILES: COC1=C(C=C(F)C=C1)C(C)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results