CAS RN 879-37-8
IUPAC Name:
2-(1H-indol-3-yl)acetamide
Synonyms:
2-(1H-indol-3-yl)acetamide
Molecular Weight (g/mol):
174.2
Molecular Formula:
C10H10N2O
InChi Key:
ZOAMBXDOGPRZLP-UHFFFAOYSA-N
SMILES:
NC(=O)CC1=CNC2=CC=CC=C12