accessibility menu, dialog, popup

UserName

CAS RN 88536-17-8

CH3OHN

CAS RN 88536-17-8

IUPAC Name: 3-ethoxypiperidine
Synonyms: 3-ethoxypiperidine
Molecular Weight (g/mol): 129.2
Molecular Formula: C7H15NO
InChi Key: RYNGVDXHFTWXDH-UHFFFAOYNA-N
SMILES: CCOC1CCCNC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results