missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxypiperidine, 96%, Thermo Scientific™

Product Code. 11429710
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11429710

Brand: Thermo Scientific Alfa Aesar H50957.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88536-17-8
Molecular Formula C7H15NO
Molecular Weight (g/mol) 129.203
MDL Number MFCD05022464
InChI Key RYNGVDXHFTWXDH-UHFFFAOYSA-N
Synonym 3-ethoxy-piperidine, piperidine, 3-ethoxy, acmc-20an4j
PubChem CID 4341745
IUPAC Name 3-ethoxypiperidine
SMILES CCOC1CCCNC1

Specifications

Quantity 1 g
Formula Weight 129.2
Percent Purity 96%
Chemical Name or Material 3-Ethoxypiperidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.