accessibility menu, dialog, popup

UserName

CAS RN 886500-13-6

IUPAC Name: 2-methoxy-4-(trifluoromethoxy)benzaldehyde
Synonyms: 2-methoxy-4-(trifluoromethoxy)benzaldehyde
Molecular Weight (g/mol): 220.15
Molecular Formula: C9H7F3O3
InChi Key: FHNKHGICKRDRLC-UHFFFAOYSA-N
SMILES: COC1=C(C=O)C=CC(OC(F)(F)F)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results