missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-(trifluoromethoxy)benzaldehyde, 97%, Thermo Scientific™

Product Code. 11465920
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465920 25 g 25g
11445920 1 g 1g
11455920 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11465920 Supplier Thermo Scientific Alfa Aesar Supplier No. H26520.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886500-13-6
Molecular Formula C9H7F3O3
Molecular Weight (g/mol) 220.147
MDL Number MFCD04115965
InChI Key FHNKHGICKRDRLC-UHFFFAOYSA-N
Synonym 2-methoxy-4-trifluoromethoxy benzaldehyde, 2-methoxy-4-trifluoromethoxy-benzaldehyde, benzaldehyde, 2-methoxy-4-trifluoromethoxy
PubChem CID 3723825
IUPAC Name 2-methoxy-4-(trifluoromethoxy)benzaldehyde
SMILES COC1=C(C=CC(=C1)OC(F)(F)F)C=O

Specifications

Quantity 25 g
Formula Weight 220.15
Percent Purity 97%
Chemical Name or Material 2-Methoxy-4-(trifluoromethoxy)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.