accessibility menu, dialog, popup

UserName

ONO

CAS RN 886851-45-2

IUPAC Name: 5-(furan-2-yl)pyridine-2-carbaldehyde
Synonyms: 5-(furan-2-yl)pyridine-2-carbaldehyde
Molecular Weight (g/mol): 173.17
Molecular Formula: C10H7NO2
InChi Key: GUIKMKZMLLANPU-UHFFFAOYSA-N
SMILES: O=CC1=NC=C(C=C1)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results