accessibility menu, dialog, popup

UserName

CAS RN 89634-74-2

ClFFFF

CAS RN 89634-74-2

IUPAC Name: 4-chloro-1-fluoro-2-(trifluoromethyl)benzene
Synonyms: 4-chloro-1-fluoro-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 198.54
Molecular Formula: C7H3ClF4
InChi Key: PFLGDQIWYTWKCM-UHFFFAOYSA-N
SMILES: FC1=C(C=C(Cl)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results