CAS RN 914295-16-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fostamatinib disodium hexahydrate, MedChemExpress
MedChemExpress Fostamatinib (R788) disodium hexahydrate is the oral prodrug of the active compound R406. R406 is an orally available and competitive Syk/FLT3 inhibitor with a Ki of 30 nM and an IC50 of 41 nM. R406 also inhibits Lyn (IC50=63 nM) and Lck (IC50=37 nM).