CAS RN 915201-07-9
IUPAC Name:
(3-chloro-5-methoxyphenyl)boronic acid
Synonyms:
3-chloro-5-methoxyphenylboronic acid
Molecular Weight (g/mol):
186.4
Molecular Formula:
C7H8BClO3
InChi Key:
WAVZSRUGKKOQRB-UHFFFAOYSA-N
SMILES:
COC1=CC(Cl)=CC(=C1)B(O)O
1
–
1
of
1
results
1
3-Chloro-5-methoxybenzeneboronic acid, 94%, Thermo Scientific™
CAS: 915201-07-9 Molecular Formula: C7H8BClO3 Molecular Weight (g/mol): 186.398 MDL Number: MFCD03095046 InChI Key: WAVZSRUGKKOQRB-UHFFFAOYSA-N Synonym: 3-chloro-5-methoxyphenyl boronic acid,3-chloro-5-methoxybenzeneboronic acid,boronic acid, 3-chloro-5-methoxyphenyl,b-3-chloro-5-methoxyphenyl-boronic acid,boronic acid, b-3-chloro-5-methoxyphenyl,pubchem1777,acmc-209rc2,3-chloro-5-methoxyphenylboronicacid,3-chloro-5-methoxyphenylboronic acid PubChem CID: 17750234 IUPAC Name: (3-chloro-5-methoxyphenyl)boronic acid SMILES: B(C1=CC(=CC(=C1)Cl)OC)(O)O