accessibility menu, dialog, popup

UserName

H3CO

CAS RN 93-18-5

IUPAC Name: 2-ethoxynaphthalene
Synonyms: bromelia
Molecular Weight (g/mol): 172.23
Molecular Formula: C12H12O
InChi Key: GUMOJENFFHZAFP-UHFFFAOYSA-N
SMILES: CCOC1=CC=C2C=CC=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results