accessibility menu, dialog, popup

UserName

ClO

CAS RN 932-67-2

IUPAC Name: cyclohex-3-ene-1-carbonyl chloride
Synonyms: cyclohex-3-ene-1-carbonyl chloride
Molecular Weight (g/mol): 144.6
Molecular Formula: C7H9ClO
InChi Key: CXWMHIRIXVNVQN-UHFFFAOYNA-N
SMILES: ClC(=O)C1CCC=CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results