accessibility menu, dialog, popup

UserName

CAS RN 934180-47-9

CH3HOFFF

CAS RN 934180-47-9

IUPAC Name: 2-methyl-3-(trifluoromethyl)phenol
Synonyms: 2-methyl-3-(trifluoromethyl)phenol
Molecular Weight (g/mol): 176.14
Molecular Formula: C8H7F3O
InChi Key: VTKJXOXHZAEQQU-UHFFFAOYSA-N
SMILES: CC1=C(C=CC=C1O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results