accessibility menu, dialog, popup

UserName

H3COOOFFF

CAS RN 93618-66-7

IUPAC Name: methyl 3-oxo-3-[3-(trifluoromethyl)phenyl]propanoate
Synonyms: methyl 3-oxo-3-[3-(trifluoromethyl)phenyl]propanoate
Molecular Weight (g/mol): 246.19
Molecular Formula: C11H9F3O3
InChi Key: RPRMYRPHNDGZOY-UHFFFAOYSA-N
SMILES: COC(=O)CC(=O)C1=CC=CC(=C1)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results