accessibility menu, dialog, popup

UserName

CAS RN 939-52-6

ClO

CAS RN 939-52-6

IUPAC Name: 4-chloro-1-phenylbutan-1-one
Synonyms: 4-chloro-1-phenylbutan-1-one
Molecular Weight (g/mol): 182.65
Molecular Formula: C10H11ClO
InChi Key: GHEFQKHLHFXSBR-UHFFFAOYSA-N
SMILES: ClCCCC(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results