accessibility menu, dialog, popup

UserName

CAS RN 944450-93-5

CH3HNSOCH3

CAS RN 944450-93-5

IUPAC Name: [(3-methoxythiophen-2-yl)methyl](methyl)amine
Synonyms: [(3-methoxythiophen-2-yl)methyl](methyl)amine
Molecular Weight (g/mol): 157.23
Molecular Formula: C7H11NOS
InChi Key: HDALHOAZSOWZLX-UHFFFAOYSA-N
SMILES: CNCC1=C(OC)C=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results