CAS RN 946135-47-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bis(dithiarsolanyl)-bis(sulfobutyl) Cyanine 3, TRC
CAS: 946135-47-3 Molecular Formula: C35H46As2N2O6S6 Molecular Weight (g/mol): 932.98 Synonym: 5-(1,3,2-Dithiarsolan-2-yl)-2-[3-[5-(1,3,2-dithiarsolan-2-yl)-1,3-dihydro-3,3-dimethyl-1-(4-sulfobutyl)-2H-indol-2-ylidene]-1-propen-1-yl]-3,3-dimethyl-1-(4-sulfobutyl)-3H-indolium Inner Salt,AsCy3; IUPAC Name: 4-[(2Z)-5-(1,3,2-dithiarsolan-2-yl)-2-[(E)-3-[5-(1,3,2-dithiarsolan-2-yl)-3,3-dimethyl-1-(4-sulfobutyl)indol-1-ium-2-yl]prop-2-enylidene]-3,3-dimethylindol-1-yl]butane-1-sulfonate SMILES: CC1(C2=C(C=CC(=C2)[As]3SCCS3)[N+](=C1/C=C/C=C/4\C(C5=C(N4CCCCS(=O)(=O)O)C=CC(=C5)[As]6SCCS6)(C)C)CCCCS(=O)(=O)[O-])C